missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ammonium pentaborate tetrahydrate, 99+%

Manufacturer:  Alfa Aesar™ 4632336

 View more versions of this product

Catalog No. AA4632336



Specifications

Specifications

Ammonium pentaborate tetrahydrate
≥99%
(NH4)2B10O16·4H2O
Ammonium borate tetrahydrate
B([O-])([O-])[O-].B([O-])([O-])[O-].B([O-])([O-])[O-].B([O-])([O-])[O-].B([O-])([O-])[O-].[NH4+].[NH4+].[NH4+].[NH4+].[NH4+].[NH4+].[NH4+].[NH4+].[NH4+].O.O.O.O
528.446
472.23 (400.17 Anhydrous)
1.58g/mL
110°C
12046-04-7
B5H44N9O19-6
MFCD13176621
OJAJAQRDJBKPPU-UHFFFAOYSA-W
nonaazanium;pentaborate;tetrahydrate
57347064
Powder
White
500g

Chemical Identifiers

12046-04-7
528.446
OJAJAQRDJBKPPU-UHFFFAOYSA-W
57347064
B([O-])([O-])[O-].B([O-])([O-])[O-].B([O-])([O-])[O-].B([O-])([O-])[O-].B([O-])([O-])[O-].[NH4+].[NH4+].[NH4+].[NH4+].[NH4+].[NH4+].[NH4+].[NH4+].[NH4+].O.O.O.O
B5H44N9O19-6
MFCD13176621
Ammonium borate tetrahydrate
nonaazanium;pentaborate;tetrahydrate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

SDS Documents