missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ammonium Oxalate TS, 3.5% (w/v) Solution, (U.S.P. Test Solution), Spectrum™

A-187, 6009-70-7

$89.69 - $331.91

Chemical Identifiers

CAS 6009-70-7
Molecular Formula C2H10N2O5
Molecular Weight (g/mol) 142.11
InChI Key MSMNVXKYCPHLLN-UHFFFAOYSA-N
IUPAC Name oxalic acid diamine hydrate
SMILES N.N.O.OC(=O)C(O)=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18600632
View Documents
Spectrum Chemical Mfg Cor
A187500MLPL
500 mL Poly Bottle
Each for $89.69
 
18600634
View Documents
Spectrum Chemical Mfg Cor
A1871LT
1 L Poly Bottle
Each for $162.89
 
18600633
View Documents
Spectrum Chemical Mfg Cor
A1874LTPL
4 L Poly Bottle
Each for $331.91
 
Description

Description

Spectrum™ Ammonium Oxalate TS, 3.5 Percent (w/v) Solution, (U.S.P. Test Solution)
Specifications

Chemical Identifiers

6009-70-7
142.11
oxalic acid diamine hydrate
C2H10N2O5
MSMNVXKYCPHLLN-UHFFFAOYSA-N
N.N.O.OC(=O)C(O)=O

Specifications

6009-70-7,7732-18-5
C2H10N2O5
N.N.O.OC(=O)C(O)=O
142.11
Poly Bottle
0.1%,99.9%
MSMNVXKYCPHLLN-UHFFFAOYSA-N
oxalic acid diamine hydrate
Chemical Solution
500 mL
SDS