missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ammonium Oxalate TS, 3.5% (w/v) Solution, (U.S.P. Test Solution), Spectrum™

A-187, 6009-70-7

Manufacturer:  Spectrum Chemical Mfg Cor A1874LTPL

 View more versions of this product

Catalog No. 18600633



Description

Description

Spectrum™ Ammonium Oxalate TS, 3.5 Percent (w/v) Solution, (U.S.P. Test Solution)
Specifications

Specifications

6009-70-7,7732-18-5
C2H10N2O5
N.N.O.OC(=O)C(O)=O
142.11
Poly Bottle
0.1%,99.9%
MSMNVXKYCPHLLN-UHFFFAOYSA-N
oxalic acid diamine hydrate
Chemical Solution
4 L

Chemical Identifiers

6009-70-7
142.11
oxalic acid diamine hydrate
C2H10N2O5
MSMNVXKYCPHLLN-UHFFFAOYSA-N
N.N.O.OC(=O)C(O)=O
SDS
Documents
Product Certifications