Please login to your online account to display your discounted pricing

Ammonium Oxalate, Saturated Solution, Certified, ≥ 5% (w/v), LabChem™

$139.11 - $139.11

Chemical Identifiers

Chemical Name or Material Ammonium Oxalate
CAS 7732-18-5
CAS Max % 95
CAS 6009-70-7
CAS Max % 5
Name Note Saturated Solution
Purity Grade Certified Grade
Linear Formula C2H8N2O4·H2O
Formula Weight 142.11
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC112822

 
lab chem
LC112822
1L Each for $139.11
Description & Specifications

Specifications

Purity Grade Certified Grade
Assay ≥5% (w/v)
Formula Weight 142.11
Chemical Name or Material Ammonium Oxalate
Identification Passes Test
Name Note Saturated Solution
Packaging Poly Bottle
Physical Form Liquid
Solubility Soluble in water
Specific Gravity 1.05g/mL
Storage Information Room Temperature
Linear Formula C2H8N2O4·H2O
CAS 7732-18-5
Decomposition Information Gaseous Ammonia; Carbon monoxide; Carbon dioxide
Color Clear, colorless
Quantity 1L
CAS Max % 95
Health Hazard 2 GHS H Statement
Solution is not hazardous.
CAS 6009-70-7
Health Hazard 3 GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
CAS Max % 5
Density 1.05g/mL
Safety and Handling

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

GHS H Statement
Solution is not hazardous.