missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ammonium oxalate monohydrate, 99+%, ACS reagent, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 423360025
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Ammonium oxalate monohydrate | |
2.5kg | |
C2H8N2O4·H2O | |
MFCD00149694 | |
ammonium oxalate monohydrate, ammonium oxalate monohydrate, acs, ethanedioic acid, diammonium salt, monohydrate, diazanium oxalate hydrate, acmc-1aw90, ksc493e5j, diammonium hydrate oxalate, oxalic acid ammonium salt monohydrate | |
MSMNVXKYCPHLLN-UHFFFAOYSA-N | |
diazanium;oxalate;hydrate | |
516808 | |
99+% | |
Plastic bottle |
6009-70-7 | |
99+% | |
(CO2NH4)2·H2O | |
15, 534 | |
Solubility in water: 45g/L (20°C). Other solubilities: slightly soluble in alcohol | |
C(=O)(C(=O)[O-])[O-].[NH4+].[NH4+].O | |
142.11 | |
142.11 | |
ACS Reagent | |
0.02% max |
Safety and Handling
GHS H Statement
Harmful in contact with skin.
Harmful if swallowed.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Call a POISON CENTER or doctor/physician if you feel unwell.
GHS Signal Word: Warning
RUO â Research Use Only