Please login to your online account to display your discounted pricing

Ammonium Oxalate, Monohydrate, ACS Grade, 99.0 to 101.0%, LabChem™

$198.09 - $198.09

Chemical Identifiers

Chemical Name or Material Ammonium Oxalate
CAS 6009-70-7
CAS Max % 100
Name Note Monohydrate
Purity Grade ACS Grade
Linear Formula C2H8N2O4·H2O
Formula Weight 142.11
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC112701

 
lab chem
LC112701
500g Each for $198.09
Description & Specifications

Specifications

Formula Weight 142.11
Chemical Name or Material Ammonium Oxalate
Identification Passes Test
Name Note Monohydrate
Packaging Poly Bottle
Purity Grade ACS Grade
Physical Form Crystals
Assay 99.0 to 101.0%
Solubility Soluble in water
Storage Information Room Temperature
Linear Formula C2H8N2O4·H2O
CAS 6009-70-7
Decomposition Information Nitrogen oxides; Carbon monoxide; Carbon dioxide; Gaseous Ammonia
Color White
Quantity 500g
Health Hazard 1 Warning
CAS Max % 100
Health Hazard 2 GHS H Statement
Harmful if swallowed or in contact with skin.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
Avoid breathing dust.
Do not eat, drink or smoke when using this product.
Use only outdoors or in a well-ventilated area.
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Call a poison center/doctor if you feel unwell.
If on skin: Wash with plenty of soap and water.
Call a poison center/doctor if you feel unwell.
Take off contaminated clothing and wash before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor if you feel unwell.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Safety and Handling

Warning

GHS P Statement
Avoid breathing dust.
Do not eat, drink or smoke when using this product.
Use only outdoors or in a well-ventilated area.
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Call a poison center/doctor if you feel unwell.
If on skin: Wash with plenty of soap and water.
Call a poison center/doctor if you feel unwell.
Take off contaminated clothing and wash before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor if you feel unwell.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Harmful if swallowed or in contact with skin.
Causes serious eye irritation.
May cause respiratory irritation.