Learn More
Ammonium oxalate monohydrate, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1026336
Description
Ammonium oxalate monohydrate is widely utilized as a buffering reagent and a dispersant to determine the surface interaction of platelets. It finds an application to study its acute poisoning effect on sheep and to investigate the formation of oxalate monoalkylamide in the human lens. It is also used in the detection and determination of calcium, lead, fluoride and rare earth metals. It is employed as chelators and forms complexes with metals. It acts as reducing agent in gold extraction and is an active ingredient of ferrous metal surface polishing fluid. It is a promoting agent in production of cobalt oxide and nickel oxide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsAmmonium oxalate monohydrate is widely utilized as a buffering reagent and a dispersant to determine the surface interaction of platelets. It finds an application to study its acute poisoning effect on sheep and to investigate the formation of oxalate monoalkylamide in the human lens. It is also used in the detection and determination of calcium, lead, fluoride and rare earth metals. It is employed as chelators and forms complexes with metals. It acts as reducing agent in gold extraction and is an active ingredient of ferrous metal surface polishing fluid. It is a promoting agent in production of cobalt oxide and nickel oxide.
Solubility
Soluble in water. Slightly soluble in alcohol.
Notes
Incompatible with strong oxidizing agents and strong acids.
Specifications
Ammonium oxalate monohydrate | |
70°C (decomposition) | |
>99% | |
(COONH4)2·H2O | |
500 g | |
3759830 | |
ammonium oxalate monohydrate, ammonium oxalate monohydrate, acs, ethanedioic acid, diammonium salt, monohydrate, diazanium oxalate hydrate, acmc-1aw90, ksc493e5j, diammonium hydrate oxalate, oxalic acid ammonium salt monohydrate | |
MSMNVXKYCPHLLN-UHFFFAOYSA-N | |
diazanium;oxalate;hydrate | |
516808 | |
98% |
6009-70-7 | |
1.5 | |
C2H10N2O5 | |
MFCD00149694 | |
UN3288 | |
14,537 | |
Soluble in water. Slightly soluble in alcohol. | |
C(=O)(C(=O)[O-])[O-].[NH4+].[NH4+].O | |
142.111 | |
142.11 (124.10 Anhydrous) |
Chemical Identifiers
6009-70-7 | |
142.111 | |
MSMNVXKYCPHLLN-UHFFFAOYSA-N | |
516808 | |
C(=O)(C(=O)[O-])[O-].[NH4+].[NH4+].O |
C2H10N2O5 | |
MFCD00149694 | |
ammonium oxalate monohydrate, ammonium oxalate monohydrate, acs, ethanedioic acid, diammonium salt, monohydrate, diazanium oxalate hydrate, acmc-1aw90, ksc493e5j, diammonium hydrate oxalate, oxalic acid ammonium salt monohydrate | |
diazanium;oxalate;hydrate |
Safety and Handling
GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.
P264b-P270-P280-P301+P312-P302+P352-P312-P330-P363-P501c
H302+H312-H320-H335-H372
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.
EINECSNumber : 238-135-4
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only