missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ammonium oxalate monohydrate, 98%, Alfa Aesar™

Manufacturer:  Alfa Aesar A102630E

 View more versions of this product

Catalog No. AAA102630E



Description

Description

Applications
Ammonium oxalate monohydrate is widely utilized as a buffering reagent and a dispersant to determine the surface interaction of platelets. It finds an application to study its acute poisoning effect on sheep and to investigate the formation of oxalate monoalkylamide in the human lens. It is also used in the detection and determination of calcium, lead, fluoride and rare earth metals. It is employed as chelators and forms complexes with metals. It acts as reducing agent in gold extraction and is an active ingredient of ferrous metal surface polishing fluid. It is a promoting agent in production of cobalt oxide and nickel oxide.

Solubility
Soluble in water. Slightly soluble in alcohol.

Notes
Incompatible with strong oxidizing agents and strong acids.
Specifications

Specifications

Ammonium oxalate monohydrate
2500g
>99%
(COONH4)2·H2O
UN3288
14,537
MSMNVXKYCPHLLN-UHFFFAOYSA-N
diazanium;oxalate;hydrate
516808
98%
70°C (decomposition)
6009-70-7
C2H10N2O5
MFCD00149694
3759830
ammonium oxalate monohydrate, ammonium oxalate monohydrate, acs, ethanedioic acid, diammonium salt, monohydrate, diazanium oxalate hydrate, acmc-1aw90, ksc493e5j, diammonium hydrate oxalate, oxalic acid ammonium salt monohydrate
C(=O)(C(=O)[O-])[O-].[NH4+].[NH4+].O
142.111
142.11 (124.10 Anhydrous)

Chemical Identifiers

6009-70-7
142.111
MSMNVXKYCPHLLN-UHFFFAOYSA-N
516808
C(=O)(C(=O)[O-])[O-].[NH4+].[NH4+].O
C2H10N2O5
MFCD00149694
ammonium oxalate monohydrate, ammonium oxalate monohydrate, acs, ethanedioic acid, diammonium salt, monohydrate, diazanium oxalate hydrate, acmc-1aw90, ksc493e5j, diammonium hydrate oxalate, oxalic acid ammonium salt monohydrate
diazanium;oxalate;hydrate
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement
P280-P301+P312-P312-P363-P322-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash contaminated clothing before reuse.
Specific measures (see label).
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.

TSCA : TSCA

SDS Documents