Please login to your online account to display your discounted pricing

Ammonium Oxalate, 3.5% (w/v), LabChem™

$112.80 - $112.80

Chemical Identifiers

Chemical Name or Material Ammonium Oxalate
CAS 7732-18-5
CAS Max % 96.5
CAS 6009-70-7
CAS Max % 3.5
Linear Formula C2H8N2O4·H2O
Formula Weight 142.11
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC112752

 
lab chem
LC112752
1L Each for $112.80
Description & Specifications

Specifications

Formula Weight 142.11
Chemical Name or Material Ammonium Oxalate
Identification Passes Test
Packaging Poly Bottle
Physical Form Liquid
Assay 3.5% (w/v)
Solubility Soluble in water
Specific Gravity 1g/mL
Storage Information Room Temperature
Linear Formula C2H8N2O4·H2O
CAS 7732-18-5
Decomposition Information Gaseous Ammonia; Carbon monoxide; Carbon dioxide
Color Clear, colorless
Quantity 1L
CAS Max % 96.5
Health Hazard 2 GHS H Statement
Solution is not hazardous.
CAS 6009-70-7
Health Hazard 3 GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
CAS Max % 3.5
Density 1g/mL
Safety and Handling

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

GHS H Statement
Solution is not hazardous.