missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ammonium Molybdate(VI) Tetrahydrate, Reagent ACS, ACROS Organics™

Manufacturer:  Fisher Scientific Acros AC423311000

 View more versions of this product

Catalog No. AC423311000


 

Specifications Description & Specifications

Specifications

1235.86
ACS Reagent Grade
White
81.0 to 83.0% (MoO3)
>150°C
50ppm max.
H24Mo7N6O24·4H2O
MFCD00167059
(NH4)6Mo7O24·4H2O
20ppm max.
200ppm max.
100ppm max.
0.001% max.
Crystalline Powder
100g
15, 530
2.4900g/mL
100ppm max.
Plastic bottle
Ammonium molybdate(VI) tetrahydrate
12054-85-2
5ppm max.
10ppm max.
0.005% max.
Passes Test (limit about 0.003%)

Chemical Identifiers

12054-85-2
(NH4)6Mo7O24·4H2O
MFCD00167059
81.0 to 83.0% (MoO3)
H24Mo7N6O24·4H2O
1235.86
Ammonium molybdate(VI) tetrahydrate
ACS Reagent Grade

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.