Please login to your online account to display your discounted pricing

Ammonium Molybdate, Tetrahydrate, ACS Grade, 81.0 to 83.0%, LabChem™

$202.40 - $202.40

Chemical Identifiers

Chemical Name or Material Ammonium Molybdate
CAS 12054-85-2
CAS Max % 100
Name Note Tetrahydrate
Purity Grade ACS Grade
Linear Formula (NH4)6Mo7O24·4H2O
Formula Weight 1235.86
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC111951

 
lab chem
LC111951
500g Each for $202.40
Description & Specifications

Specifications

Formula Weight 1235.86
Chemical Name or Material Ammonium Molybdate
Identification Passes Test
Name Note Tetrahydrate
Packaging Poly Bottle
Purity Grade ACS Grade
Physical Form Crystals
Assay 81.0 to 83.0%
Assay Percent Notes as MoO3
Solubility Soluble in water
Specific Gravity 2.276g/cm3
Storage Information Room Temperature
Linear Formula (NH4)6Mo7O24·4H2O
CAS 12054-85-2
Decomposition Information Gaseous Ammonia
Color White
Quantity 500g
Health Hazard 1 Warning
CAS Max % 100
Health Hazard 2 GHS H Statement
Causes skin irritation and serious eye irritation.
May cause respiratory irritation.
Harmful to aquatic life.
Health Hazard 3 GHS P Statement
Avoid breathing dust.
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
Use only outdoors or in a well-ventilated area.
Avoid release to the environment.
If on skin: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
Take off contaminated clothing and wash before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor if you feel unwell.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Store locked up in a well-ventilated place.
Keep container tightly closed.
Dispose of contents/container to comply with local, state and federal regulations.
Density 2.276g/cm3
Safety and Handling

Warning

GHS P Statement
Avoid breathing dust.
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
Use only outdoors or in a well-ventilated area.
Avoid release to the environment.
If on skin: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
Take off contaminated clothing and wash before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor if you feel unwell.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Store locked up in a well-ventilated place.
Keep container tightly closed.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Causes skin irritation and serious eye irritation.
May cause respiratory irritation.
Harmful to aquatic life.