Please login to your online account to display your discounted pricing

Ammonium Molybdate, For Silica, Certified, 7.5% (w/v), LabChem™

$60.54 - $60.54

Chemical Identifiers

Chemical Name or Material Ammonium Molybdate
CAS 7732-18-5
CAS Max % 92.5
CAS 12054-85-2
CAS Max % 7.5
Name Note For Silica
Purity Grade Certified Grade
Linear Formula (NH4)6Mo7O24·4H2O
Formula Weight 1235.86
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC112201

 
lab chem
LC112201
500mL Each for $60.54
Description & Specifications

Specifications

Formula Weight 1235.86
Purity Grade Certified Grade
Solubility Soluble in water
Quantity 500mL
CAS 7732-18-5
Suitability Passes Test
Chemical Name or Material Ammonium Molybdate
Health Hazard 3 GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
CAS Max % 92.5
Density 1g/mL
Identification Passes Test
CAS Max % 7.5
Assay 7.5% (w/v)
Health Hazard 2 GHS H Statement
Solution is not hazardous.
Specific Gravity 1g/mL
Storage Information Room Temperature
Decomposition Information Gaseous Ammonia
Color Clear, colorless
Physical Form Liquid
Linear Formula (NH4)6Mo7O24·4H2O
Name Note For Silica
CAS 12054-85-2
Packaging Poly Bottle
Safety and Handling

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

GHS H Statement
Solution is not hazardous.