missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ammonium Molybdate, For Silica, Certified, 10% (w/v), LabChem™

Manufacturer:  LabChem LC112504

 View more versions of this product

Catalog No. LC112504



Specifications

Specifications

Ammonium Molybdate
7732-18-5,12054-85-2,1310-73-2
(NH4)6Mo7O24·4H2O
water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide
Liquid
Certified
Poly Bottle
1.02g/mL
Room Temperature
Gaseous Ammonia
4L
For Silica
87.91,9.69,2.4
1.02g/mL
1235.86
10% (w/v)
Passes Test
Soluble in water
Passes Test
1.1cSt
Colorless

Chemical Identifiers

7732-18-5
water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing.
If skin irritation occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.

Warning

Documents
Promotions

Promotions