Please login to your online account to display your discounted pricing

Ammonium Molybdate, For Phosphorus, Certified, 4% (w/v), LabChem™

$91.69 - $91.69

Chemical Identifiers

Chemical Name or Material Ammonium Molybdate
CAS 7732-18-5
CAS Max % 96
CAS 12054-85-2
CAS Max % 4
Name Note For Phosphorus
Purity Grade Certified Grade
Linear Formula (NH4)6Mo7O24·4H2O
Formula Weight 1235.86
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC112101

 
lab chem
LC112101
500mL Each for $91.69
Description & Specifications

Specifications

Purity Grade Certified Grade
Formula Weight 1235.86
Chemical Name or Material Ammonium Molybdate
CAS Max % 4
Assay 4% (w/v)
Density 1g/mL
Suitability Passes Test
Quantity 500mL
Health Hazard 2 GHS H Statement
Solution is not hazardous.
CAS 12054-85-2
Packaging Poly Bottle
Storage Information Room Temperature
CAS Max % 96
Identification Passes Test
Specific Gravity 1g/mL
Decomposition Information Gaseous Ammonia
Solubility Soluble in water
CAS 7732-18-5
Linear Formula (NH4)6Mo7O24·4H2O
Name Note For Phosphorus
Physical Form Liquid
Health Hazard 3 GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Color Clear, colorless
Safety and Handling

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

GHS H Statement
Solution is not hazardous.