Please login to your online account to display your discounted pricing

Ammonium Molybdate, For Phosphorus, Certified, 4.8% (w/v), LabChem™

$92.34 - $92.34

Chemical Identifiers

Chemical Name or Material Ammonium Molybdate
CAS 7732-18-5
CAS Max % 94.95
CAS 12054-85-2
CAS Max % 4.8
CAS 1336-21-6
CAS Max % 0.25
Name Note For Phosphorus
Purity Grade Certified Grade
Linear Formula (NH4)6Mo7O24·4H2O
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC112151

 
lab chem
LC112151
500mL Each for $92.34
Description & Specifications

Specifications

Purity Grade Certified Grade
Formula Weight 1235.86
Chemical Name or Material Ammonium Molybdate
Color Clear, colorless
Assay 4.8% (w/v)
Linear Formula (NH4)6Mo7O24·4H2O
CAS 12054-85-2
Packaging Poly Bottle
CAS Max % 4.8
CAS Max % 0.25
Decomposition Information Gaseous Ammonia
Quantity 500mL
Name Note For Phosphorus
Physical Form Liquid
Density 1g/mL
CAS 7732-18-5
CAS Max % 94.95
Health Hazard 2 GHS H Statement
Solution is not hazardous.
Identification Passes Test
Health Hazard 3 GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Solubility Soluble in water
CAS 1336-21-6
Specific Gravity 1g/mL
Suitability Passes Test
Storage Information Room Temperature
Safety and Handling

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

GHS H Statement
Solution is not hazardous.