missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ammonium molybdate (di), Mo 56.5%

Manufacturer:  Alfa Aesar™ 1221518*

 View more versions of this product

Catalog No. AA1221518



Specifications

Specifications

Ammonium molybdate (di)
H2MoO4
Ammonium dimolybdate
161.96
Powder
50g
27546-07-2
MFCD00010882
VLAPMBHFAWRUQP-UHFFFAOYSA-L
339.95
Mo 56.5%

Chemical Identifiers

27546-07-2
161.96
VLAPMBHFAWRUQP-UHFFFAOYSA-L
H2MoO4
MFCD00010882
Ammonium dimolybdate
Safety and Handling

Safety and Handling

TSCA : TSCA

Documents