Please login to your online account to display your discounted pricing

Ammonium Molybdate ACS AR Crystal, Macron Fine Chemicals™

macron

Chemical Identifiers

Chemical Name or Material Ammonium Molybdate
Name Note 4-Hydrate
CAS 12054-85-2
Purity Grade ACS
Physical Form Crystalline
Molecular Formula (NH4)6Mo7O24·4H2O
Formula Weight 1235.86g/mol
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MK3420125

 
avantor performance
342002
POLYSTORMOR 125g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Packaging POLYSTORMOR
Quantity 125g
Chemical Name or Material Ammonium Molybdate
Name Note 4-Hydrate
CAS 12054-85-2
Purity Grade ACS
Physical Form Crystalline
Molecular Formula (NH4)6Mo7O24·4H2O
Formula Weight 1235.86g/mol