missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ammonium Molybdate, 1% (w/v) in 0.1N Sulfuric Acid, Certified, LabChem™

$89.97 - $89.97

Chemical Identifiers

CAS 7732-18-5,12054-85-2,7664-93-9
CAS Max % 98.51,1,0.49
Linear Formula (NH4)6Mo7O24·4H2O
Formula Weight 1235.86
Chemical Name or Material Ammonium Molybdate, 1% (w/v) in 0.1N Sulfuric Acid
Grade Certified Grade
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
LC112052 Lab Chem
LC112052
1L Each for $89.97
Specifications Description & Specifications

Chemical Identifiers

7732-18-5,12054-85-2,7664-93-9
(NH4)6Mo7O24·4H2O
Ammonium Molybdate, 1% (w/v) in 0.1N Sulfuric Acid
98.51,1,0.49
1235.86
Certified Grade

Specifications

7732-18-5,12054-85-2,7664-93-9
Liquid
1235.86
Passes Test
Soluble in water
Room Temperature
1.01cSt
Clear, colorless
Ammonium Molybdate, 1% (w/v) in 0.1N Sulfuric Acid
98.51,1,0.49
(NH4)6Mo7O24·4H2O
1g/mL
Poly Bottle
1g/mL
Passes Test
Sulfur compounds
1L
Certified Grade

Safety and Handling

GHS H Statement
Solution is not hazardous.

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.