Please login to your online account to display your discounted pricing

Ammonium Metavanadate, (White to light yellow powder),MP Biomedicals

Cell Culture Reagent

$63.00 - $65.77

Chemical Identifiers

Chemical Name or Material Ammonium Metavanadate
CAS 7803-55-6
Physical Form Powder
Molecular Formula NH4VO3
Formula Weight 117
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

ICN15477580

 
mp biomedicals
15477580
100g Each for $63.00

ICN19462480

 
mp biomedicals
19462480
100g Each for $65.77
Description & Specifications

Specifications

Physical Form Powder
Formula Weight 117
Color White to light yellow
Quantity 100g
Chemical Name or Material Ammonium Metavanadate
Molecular Formula NH4VO3
CAS 7803-55-6
Recommended Storage Room Temperature
Safety and Handling

Room Temperature