missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Honeywell Fluka™ Ammonium Metavanadate, puriss. p.a. ACS Reagent, ≥99.0% (RT), Honeywell™ Fluka™

Puriss. p.a., ACS reagent,≥99.0% (RT)

$426.92 - $426.92

Chemical Identifiers

CAS 7803-55-6
Molecular Formula H4NO3V
Molecular Weight (g/mol) 116.977
MDL Number MFCD00011430
InChI Key UNTBPXHCXVWYOI-UHFFFAOYSA-O
Synonym ammonium metavanadate, ammonium vanadate v, ammonium monovanadate, unii-fl85px638g, vanadate vo31-, ammonium, vanadate vo31-, ammonium 1:1, ammonium vanadium oxide, ammoniummetavanadate, vanadic acid, hvo3 , ammonium salt, ammonium meta-vanadate
PubChem CID 516859
IUPAC Name azanium;oxido(dioxo)vanadium
SMILES [NH4+].[O-][V](=O)=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
60044634 View Documents Honeywell Fluka™
10028500G
500g Poly Bottle Each for $426.92
 
Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Chemical Identifiers

7803-55-6
116.977
UNTBPXHCXVWYOI-UHFFFAOYSA-O
516859
[NH4+].[O-][V](=O)=O
H4NO3V
MFCD00011430
ammonium metavanadate, ammonium vanadate v, ammonium monovanadate, unii-fl85px638g, vanadate vo31-, ammonium, vanadate vo31-, ammonium 1:1, ammonium vanadium oxide, ammoniummetavanadate, vanadic acid, hvo3 , ammonium salt, ammonium meta-vanadate
azanium;oxido(dioxo)vanadium

Specifications

7803-55-6
NH4VO3
UN2859
ammonium metavanadate, ammonium vanadate v, ammonium monovanadate, unii-fl85px638g, vanadate vo31-, ammonium, vanadate vo31-, ammonium 1:1, ammonium vanadium oxide, ammoniummetavanadate, vanadic acid, hvo3 , ammonium salt, ammonium meta-vanadate
[NH4+].[O-][V](=O)=O
116.977
116.98g/mol
ACS/puriss. p.a.
Poly Bottle
200°C
500g
H4NO3V
MFCD00011430
Powder/Crystals
UNTBPXHCXVWYOI-UHFFFAOYSA-O
azanium;oxido(dioxo)vanadium
516859
≥99.0% (RT)
2.32g/cu.cm at 25°C
Beige-Yellow to White
7 (at 5.10g/L)
Ammonium Metavanadate
Safety and Handling

Safety and Handling

P260-P284-P301 + P310-P305 + P351 + P338-P310

H301-H315-H319-H330-H335

DOTInformation : 6.1/PGII

EINECSNumber : 232-261-3

RTECSNumber : YW0875000