missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ammonium metavanadate, 99.5%, for analysis, ACROS Organics™

Manufacturer:  Acros Organics AC194910010

 View more versions of this product

Catalog No. AC194910010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Ammonium metavanadate
99.5%
NH4VO3
UNTBPXHCXVWYOI-UHFFFAOYSA-O
azanium;oxido(dioxo)vanadium
516859
Crystalline Powder
Analysis
20ppm max.
10ppm max.
20ppm max.
Glass bottle
Solubility in water: 5.1g/L (20°C). Other solubilities: insoluble in ethanol, 69.5g/L (96°C)
10ppm max.
200.0°C
7803-55-6
H4NO3V
ammonium meta-vanadate
[NH4+].[O-][V](=O)=O
116.977
116.98
99.5%
10ppm max.
20ppm max.
10ppm max.
20ppm max.
50ppm max.
100ppm max.
Green to White
1kg

Chemical Identifiers

7803-55-6
116.977
ammonium meta-vanadate
azanium;oxido(dioxo)vanadium
H4NO3V
UNTBPXHCXVWYOI-UHFFFAOYSA-O
516859
[NH4+].[O-][V](=O)=O
Safety and Handling

Safety and Handling

Danger

SDS Documents