Please login to your online account to display your discounted pricing

Ammonium meta-Vanadate, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™


High quality chemicals for laboratory and specialized industrial use

Quantity 125g
Packaging Wide Mouth Amber Glass Bottle

Manufacturer: avantor performance  071504

 View more versions of this product

Catalog No. 02002302

  • / Each
  • / Case of 4 Each
 

Description & Specifications

Chemical Identifiers

Purity Grade Baker Analyzed™ ACS Reagent
Formula Weight 116.98
Chemical Name or Material Ammonium meta-Vanadate
Molecular Formula NH4VO3
CAS 7803-55-6
Packaging Wide Mouth Amber Glass Bottle
Specific Gravity 1L = 2.33kg
Quantity 125g

Specifications

Chemical Name or Material Ammonium meta-Vanadate
CAS 7803-55-6
Purity Grade Baker Analyzed™ ACS Reagent
Molecular Formula NH4VO3
Formula Weight 116.98