Please login to your online account to display your discounted pricing

Ammonium meta-Vanadate, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$594.52 - $879.90

Chemical Identifiers

Chemical Name or Material Ammonium meta-Vanadate
CAS 7803-55-6
Purity Grade Baker Analyzed™ ACS Reagent
Molecular Formula NH4VO3
Formula Weight 116.98
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02002303

 
avantor performance
071501
Wide Mouth Amber Glass Bottle 500g Each for $594.52

02002302

 
avantor performance
071504
Wide Mouth Amber Glass Bottle 125g Each for $879.90
Case of 4 Each for $3,519.60
Description & Specifications

Specifications

Purity Grade Baker Analyzed™ ACS Reagent
Formula Weight 116.98
Chemical Name or Material Ammonium meta-Vanadate
CAS 7803-55-6
Packaging Wide Mouth Amber Glass Bottle
Specific Gravity 1L = 2.33kg
Quantity 500g
Molecular Formula NH4VO3