Learn More
Ammonium magnesium phosphate hexahydrate, 98%, Thermo Scientific Chemicals
$152.48 - $504.42
Chemical Identifiers
CAS | 13478-16-5 |
---|---|
Molecular Formula | H16MgNO10P |
Molecular Weight (g/mol) | 245.404 |
MDL Number | MFCD00150461 |
InChI Key | CKMXBZGNNVIXHC-UHFFFAOYSA-L |
Synonym | unii-aw3ejl1462, magnesium ammonium phosphate hexahydrate, acmc-20alsk, magnesium 2+ ion ammonium hexahydrate phosphate, magnesium 2+ ammonium hexahydrate phosphate, 7785-21-9 h3po4-ammonium-magnesium 1:1:1 |
PubChem CID | 10220511 |
IUPAC Name | azanium;magnesium;phosphate;hexahydrate |
SMILES | [NH4+].O.O.O.O.O.O.[O-]P(=O)([O-])[O-].[Mg+2] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1749722
|
Thermo Scientific Chemicals
A1749722 |
100 g |
N/A
|
|
|||||
AAA1749736
|
Thermo Scientific Chemicals
A1749736 |
500 g |
N/A
|
|
|||||
Description
It is used as pharmaceutical intermediae.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used as pharmaceutical intermediae.
Solubility
Insoluble in water.
Notes
Store in a cool place. Ensure proper ventilation.
Chemical Identifiers
13478-16-5 | |
245.404 | |
CKMXBZGNNVIXHC-UHFFFAOYSA-L | |
10220511 | |
[NH4+].O.O.O.O.O.O.[O-]P(=O)([O-])[O-].[Mg+2] |
H16MgNO10P | |
MFCD00150461 | |
unii-aw3ejl1462, magnesium ammonium phosphate hexahydrate, acmc-20alsk, magnesium 2+ ion ammonium hexahydrate phosphate, magnesium 2+ ammonium hexahydrate phosphate, 7785-21-9 h3po4-ammonium-magnesium 1:1:1 | |
azanium;magnesium;phosphate;hexahydrate |
Specifications
13478-16-5 | |
H16MgNO10P | |
unii-aw3ejl1462, magnesium ammonium phosphate hexahydrate, acmc-20alsk, magnesium 2+ ion ammonium hexahydrate phosphate, magnesium 2+ ammonium hexahydrate phosphate, 7785-21-9 h3po4-ammonium-magnesium 1:1:1 | |
CKMXBZGNNVIXHC-UHFFFAOYSA-L | |
azanium;magnesium;phosphate;hexahydrate | |
10220511 | |
98% | |
Ammonium magnesium phosphate hexahydrate |
100 g | |
MFCD00150461 | |
Insoluble in water. | |
[NH4+].O.O.O.O.O.O.[O-]P(=O)([O-])[O-].[Mg+2] | |
245.404 | |
245.41 (137.32 Anhydrous) | |
1.711 |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 232-075-2
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only