Learn More
Ammonium L-(+)-tartrate, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 017658A1
Description
Ammonium L-(+)-tartrate finds application in the textile and pharmaceutical industries. It is also used to study protein structural analysis, biochemicals and reagents, biological buffers, chelators and proteomics. It acts as a nitrogen source in biological applications and cell culture media. Further, it is used in the study of crystal optimization research and preliminary diffraction data analysis of the Smad1 MH1 domain bonded to a palindromic SBE DNA element.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Ammonium-L-tartrate | |
Crystalline | |
C4H12N2O6 | |
6120352 | |
Soluble in water and alcohol. | |
N.N.O[C@H]([C@@H](O)C(O)=O)C(O)=O | |
184.15 | |
CHEBI:63075 | |
Hygroscopic |
3164-29-2 | |
1 kg | |
MFCD00013073 | |
ammonium d-tartrate, diammonium l-+-tartrate, l-tartaric acid, ammonium salt, unii-ji8ud88354, diammonium l-tartrate, r-2,3-dihydroxybutanedioic acid diammonium salt, ammonium tartrate dibasic, butanedioic acid, 2,3-dihydroxy-2r,3r-, diammonium salt, ammonium l-+-tartrate, diammonium 2r,3r-2,3-dihydroxysuccinate | |
NGPGDYLVALNKEG-OLXYHTOASA-N | |
(2R,3R)-2,3-dihydroxybutanedioic acid diamine | |
13652278 | |
184.15 | |
1.6 |
Chemical Identifiers
3164-29-2 | |
184.15 | |
NGPGDYLVALNKEG-OLXYHTOASA-N | |
13652278 | |
(2R,3R)-2,3-dihydroxybutanedioic acid diamine |
C4H12N2O6 | |
MFCD00013073 | |
ammonium d-tartrate, diammonium l-+-tartrate, l-tartaric acid, ammonium salt, unii-ji8ud88354, diammonium l-tartrate, r-2,3-dihydroxybutanedioic acid diammonium salt, ammonium tartrate dibasic, butanedioic acid, 2,3-dihydroxy-2r,3r-, diammonium salt, ammonium l-+-tartrate, diammonium 2r,3r-2,3-dihydroxysuccinate | |
CHEBI:63075 | |
N.N.O[C@H]([C@@H](O)C(O)=O)C(O)=O |
Safety and Handling
EINECSNumber : 221-618-9
RTECSNumber : WW8050000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only