Please login to your online account to display your discounted pricing

Ammonium L-(+)-tartrate, 98%, Alfa Aesar™

$27.73 - $27.73

Chemical Identifiers

Chemical Name or Material Ammonium L-(+)-tartrate
CAS 3164-29-2
Molecular Formula C4H12N2O6
Formula Weight 184.15
MDL Number MFCD00013073
Beilstein 6120352
Synonym L-(+)-Tartaric acid diammonium salt; Ammonium tartrate, dibasic
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AA1765818

  SDS
alfa aesar
1765818
Each for $27.73
Description & Specifications

Specifications

Formula Weight 184.15
Quantity 50g
Beilstein 6120352
Chemical Name or Material Ammonium L-(+)-tartrate
Density 1.6
Molecular Formula C4H12N2O6
Synonym L-(+)-Tartaric acid diammonium salt; Ammonium tartrate, dibasic
MDL Number MFCD00013073
Sensitivity Hygroscopic
Odor Odorless
Solubility Information Soluble in water, alcohol
CAS 3164-29-2
Assay 98%
Color White
RTECS Number WW8050000
Physical Form Crystalline
TSCA TSCA