missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ammonium iron(III) sulfate dodecahydrate, MilliporeSigma™

For analysis EMSURE™ ACS, ISO, Reag. Ph Eur

Manufacturer:  Emd Millipore Corporation 1037769050

 View more versions of this product

Catalog No. M1037769050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

7783-83-7
ammonium ferric sulfate, ammonium iron iii sulfate dodecahydrate, acs 100g, azanium iron +3 cation disulfate dodecahydrate, azanium iron 3+ disulfate dodecahydrate, ferric ammonium sulfate dodecahydrate, ferric ammonium sulfate, acs, ferric iron ammonium dodecahydrate disulfate, iron ammonium sulfate dodecahydrate, sulfuric acid, ammonium iron 3+ salt 2:1:1 , dodecahydrate
LCPUDZUWZDSKMX-UHFFFAOYSA-K
azanium;iron(3+);disulfate;dodecahydrate
197097
Odorless
ACS, ISO, Reag. Ph Eur
FeH28NO20S2
Soluble in water: 1240g/L at 25°C
[NH4+].O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Fe+3]
482.176
482.19
Fibre carton
50kg

Chemical Identifiers

7783-83-7
482.176
ammonium ferric sulfate, ammonium iron iii sulfate dodecahydrate, acs 100g, azanium iron +3 cation disulfate dodecahydrate, azanium iron 3+ disulfate dodecahydrate, ferric ammonium sulfate dodecahydrate, ferric ammonium sulfate, acs, ferric iron ammonium dodecahydrate disulfate, iron ammonium sulfate dodecahydrate, sulfuric acid, ammonium iron 3+ salt 2:1:1 , dodecahydrate
azanium;iron(3+);disulfate;dodecahydrate
FeH28NO20S2
LCPUDZUWZDSKMX-UHFFFAOYSA-K
197097
[NH4+].O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Fe+3]
Safety and Handling

Safety and Handling

EINECSNumber : 233-382-4

Recommended Storage : Store below 15°C (59°F), Tightly closed. Dry. Protected from Light.

Documents