Learn More
Ammonium iron(III) sulfate dodecahydrate, 99+%, for analysis, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 205880500
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
230.0°C | |
39.0°C to 41.0°C | |
7783-83-7 | |
Adhering Crystalline Powder or Crystals | |
99% min. (Iodometry) | |
(NH4)Fe(SO4)2·12H2O | |
15, 515 | |
Solubility in water: 1240g/L water (25°C). Other solubilities: practically insoluble in alcohol | |
[NH4+].O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Fe+3] | |
482.19 | |
482.19 | |
Analytical | |
1.71 |
Ammonium iron(III) sulfate dodecahydrate | |
p.a. | |
Gray-Purple | |
50 g | |
H4FeNO8S2·12H2O | |
MFCD00150004 | |
ammonium ferric sulfate, ferric ammonium sulfate dodecahydrate, iron ammonium sulfate dodecahydrate, sulfuric acid, ammonium iron 3+ salt 2:1:1 , dodecahydrate, ferric ammonium sulfate, acs, azanium iron 3+ disulfate dodecahydrate, ferric iron ammonium dodecahydrate disulfate, azanium iron +3 cation disulfate dodecahydrate, ammonium iron iii sulfate dodecahydrate, acs 100g | |
LCPUDZUWZDSKMX-UHFFFAOYSA-K | |
azanium;iron(3+);disulfate;dodecahydrate | |
197097 | |
≥99% | |
Plastic bottle | |
1.7100g/mL |
Chemical Identifiers
7783-83-7 | |
482.19 | |
LCPUDZUWZDSKMX-UHFFFAOYSA-K | |
197097 | |
[NH4+].O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Fe+3] |
H4FeNO8S2·12H2O | |
MFCD00150004 | |
ammonium ferric sulfate, ferric ammonium sulfate dodecahydrate, iron ammonium sulfate dodecahydrate, sulfuric acid, ammonium iron 3+ salt 2:1:1 , dodecahydrate, ferric ammonium sulfate, acs, azanium iron 3+ disulfate dodecahydrate, ferric iron ammonium dodecahydrate disulfate, azanium iron +3 cation disulfate dodecahydrate, ammonium iron iii sulfate dodecahydrate, acs 100g | |
azanium;iron(3+);disulfate;dodecahydrate |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Wear protective gloves/protective clothing/eye protec
GHS Signal Word: Warning
RTECSNumber : WS5900000
RUO – Research Use Only