Please login to your online account to display your discounted pricing

Ammonium iron(III) oxalate trihydrate, 98%, pure, ACROS Organics™


Quantity 1kg
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC335100010

 View more versions of this product

Catalog No. AC335100010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Crystalline Powder
Formula Weight 428.07
Quantity 1kg
Color Green
Purity Grade Pure
Linear Formula (NH4)3Fe(C2O4)3·3H2O
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Molecular Formula C6H12FeN3O12·3H2O
Chemical Name or Material Ammonium iron(III) oxalate trihydrate
Assay Percent Range 98%
MDL Number MFCD00150137
CAS 13268-42-3
Packaging Glass bottle
Infrared Spectrum Authentic
Synonym Ammonium ferric oxalate
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Health Hazard 1 GHS Signal Word: Danger

Specifications

Chemical Name or Material Ammonium iron(III) oxalate trihydrate
CAS 13268-42-3
Purity Grade Pure
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C6H12FeN3O12·3H2O
Linear Formula (NH4)3Fe(C2O4)3·3H2O
Formula Weight 428.07
MDL Number MFCD00150137
Synonym Ammonium ferric oxalate
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

SDS & Safety Documents