missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ammonium iron(III) oxalate trihydrate, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B2141122

 View more versions of this product

Catalog No. AAB2141122



Description

Description

Ammonium iron(III) oxalate trihydrate is used in the preparation of Langmuir-Blodgett magnetic thin films. It is also involved in the production of blueprint paper.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ammonium iron(III) oxalate trihydrate is used in the preparation of Langmuir-Blodgett magnetic thin films. It is also involved in the production of blueprint paper.

Solubility
Soluble in water. Insoluble in ethanol.

Notes
Light sensitive and hygroscopic. Incompatible with strong oxidizing agents and strong acids.
Specifications

Specifications

Ammonium iron(III) oxalate trihydrate
100 g
MFCD00150137
14,517
Soluble in water. Insoluble in ethanol.
[NH4+].[NH4+].[NH4+].O.O.O.O=C1O[Fe+3]234(OC1=O)OC(=O)C(=O)O2.O=C(O3)C(=O)O4
428.06
98%
1.78
13268-42-3
C6H18FeN3O15
UN3288
Ferric ammonium oxalate; Iron(III) ammonium oxalate
ARJSTCBPHIPXIJ-UHFFFAOYSA-K
triammonium hexaoxo-1,4,6,9,10,13-hexaoxa-5-ferraspiro[4.4⁵.4⁵]tridecane-5,5,5-tris(ylium) trihydrate
428.07 (374.02 Anhydrous)
Hygroscopic; Light sensitive

Chemical Identifiers

13268-42-3
428.06
ARJSTCBPHIPXIJ-UHFFFAOYSA-K
triammonium hexaoxo-1,4,6,9,10,13-hexaoxa-5-ferraspiro[4.4⁵.4⁵]tridecane-5,5,5-tris(ylium) trihydrate
C6H18FeN3O15
MFCD00150137
Ferric ammonium oxalate; Iron(III) ammonium oxalate
[NH4+].[NH4+].[NH4+].O.O.O.O=C1O[Fe+3]234(OC1=O)OC(=O)C(=O)O2.O=C(O3)C(=O)O4
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.

P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H302-H312-H314-H335

DOTInformation : Hazard Class: 8; Packaging Group: III

EINECSNumber : 238-090-0

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only