Please login to your online account to display your discounted pricing

Alfa Aesar™ Ammonium iron(III) oxalate hydrate

In photography, blueprints

Chemical Identifiers

Chemical Name or Material Ammonium iron(III) oxalate hydrate
CAS 13268-42-3
Formula Weight 374.02 (Anhydrous)
MDL Number MFCD00013071
UN Number UN3288
Merck Index 14,517
Synonym Ferric ammonium oxalate; Iron(III) ammonium oxalate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4211218

  SDS
alfa aesar™
4211218
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4211230

  SDS
alfa aesar™
4211230
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 374.02 (Anhydrous)
Chemical Name or Material Ammonium iron(III) oxalate hydrate
TSCA TSCA
Physical Form Powder
MDL Number MFCD00013071
CAS 13268-42-3
UN Number UN3288
Quantity 50g
Merck Index 14,517
Sensitivity Light Sensitive & Hygroscopic
Synonym Ferric ammonium oxalate; Iron(III) ammonium oxalate
Solubility Information Very soluble in water; Insoluble in alcohol
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.
Health Hazard 3 GHS P Statement
P280-P301+P312-P312-P363-P322-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash contaminated clothing before reuse.
Specific measures (see label).
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.
Safety and Handling

Warning

GHS P Statement
P280-P301+P312-P312-P363-P322-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash contaminated clothing before reuse.
Specific measures (see label).
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.