missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ammonium iron(III) hexacyanoferrate(II) hydrate, tech., Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2190822
Description
It is employed as a intermediate for pharmaceuticals. Also used in decorative cosmetics.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is employed as a intermediate for pharmaceuticals. Also used in decorative cosmetics.
Solubility
Soluble in water and mineral acids. Insoluble in alkali.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications
Ammonium iron(III) hexacyanoferrate(II) hydrate | |
100 g | |
MFCD01726733 | |
ammonium iron iii hexacyanoferrate ii, iron 3+ ammonium ferrocyanide, azanium;iron 2+ ;iron 3+ ;hexacyanide, ammonium iron iii hexacyanoferrate ii hydrate, ammonium iron iii hexacyanoferrate ii hydrate, tech., ammonium iron iii hexacyanoferrate ii giese salt | |
ILZWGESBVHGTRX-UHFFFAOYSA-O | |
azanium;iron(2+);iron(3+);hexacyanide | |
23620040 | |
Technical |
25869-00-5 | |
C6H4Fe2N7 | |
14,7910 | |
Soluble in water and mineral acids. Insoluble in alkali. | |
[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[NH4+].[Fe+2].[Fe+3] | |
285.837 | |
285.83 (Anhydrous) |
Chemical Identifiers
25869-00-5 | |
285.837 | |
ILZWGESBVHGTRX-UHFFFAOYSA-O | |
23620040 | |
[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[NH4+].[Fe+2].[Fe+3] |
C6H4Fe2N7 | |
MFCD01726733 | |
ammonium iron iii hexacyanoferrate ii, iron 3+ ammonium ferrocyanide, azanium;iron 2+ ;iron 3+ ;hexacyanide, ammonium iron iii hexacyanoferrate ii hydrate, ammonium iron iii hexacyanoferrate ii hydrate, tech., ammonium iron iii hexacyanoferrate ii giese salt | |
azanium;iron(2+);iron(3+);hexacyanide |
Safety and Handling
EINECSNumber : 247-304-1
RTECSNumber : LJ8140000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only