missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Honeywell™ Ammonium Iron(III) Citrate, Reagent Grade, Honeywell™

Reagent Grade, powder

$151.79 - $151.79

Chemical Identifiers

CAS 1185-57-5
Molecular Formula C6H11FeNO7+3
Molecular Weight (g/mol) 264.999
InChI Key FRHBOQMZUOWXQL-UHFFFAOYSA-N
Synonym ammonium ferric citrate, ferri seltz, ammonii ferri citras, ferric ammonium citrate, iron ammonium citrate, ammonium iron 3+ citrate, ammonium iron iii citrate, iron ammonium citrate, green, ferri-ammoniumcitrat, braunes, ferriseltz
PubChem CID 118984355
IUPAC Name azane;2-hydroxypropane-1,2,3-tricarboxylic acid;iron(3+)
SMILES C(C(=O)O)C(CC(=O)O)(C(=O)O)O.N.[Fe+3]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
60048007 View Documents Honeywell Chemicals
F5879100G
100g Poly bottle Each for $151.79
 
Description

Description

  • ≥95% for general laboratory use.
Specifications

Chemical Identifiers

1185-57-5
264.999
ammonium ferric citrate, ferri seltz, ammonii ferri citras, ferric ammonium citrate, iron ammonium citrate, ammonium iron 3+ citrate, ammonium iron iii citrate, iron ammonium citrate, green, ferri-ammoniumcitrat, braunes, ferriseltz
azane;2-hydroxypropane-1,2,3-tricarboxylic acid;iron(3+)
C6H11FeNO7+3
FRHBOQMZUOWXQL-UHFFFAOYSA-N
118984355
C(C(=O)O)C(CC(=O)O)(C(=O)O)O.N.[Fe+3]

Specifications

1185-57-5
C6H8O7 ·xFe3+ · yNH3
FRHBOQMZUOWXQL-UHFFFAOYSA-N
azane;2-hydroxypropane-1,2,3-tricarboxylic acid;iron(3+)
118984355
Solid
100g
C6H11FeNO7+3
ammonium ferric citrate, ferri seltz, ammonii ferri citras, ferric ammonium citrate, iron ammonium citrate, ammonium iron 3+ citrate, ammonium iron iii citrate, iron ammonium citrate, green, ferri-ammoniumcitrat, braunes, ferriseltz
C(C(=O)O)C(CC(=O)O)(C(=O)O)O.N.[Fe+3]
264.999
Poly bottle
Green
Ammonium iron(III) citrate
Safety and Handling

Safety and Handling

P261-P305 + P351 + P338

RTECSNumber : 214-686-6