Learn More
Ammonium iron(III) citrate, brown
CAS: 1185-57-5 | C6H11FeNO7+3 | 264.999 g/mol
Supplier: Thermo Scientific Chemicals A111990B
Description
Ammonium iron(III) citrate, brown is used in medicine, in making blueprints, and as a feed additive. It is also used as an iron supplement and food enhancer.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsAmmonium iron(III) citrate, brown is used in medicine, in making blueprints, and as a feed additive. It is also used as an iron supplement and food enhancer.
Solubility
Soluble in water.1200 g/L (20°C)
Notes
Hygroscopic. Desiccate at room temperature. Store in the dark. Store under inert gas.
Specifications
| Ammonium iron (III) citrate | |
| 1185-57-5 | |
| C6H11FeNO7+3 | |
| 14,4017 | |
| Soluble in water.1200g/L (20°C) | |
| C(C(=O)O)C(CC(=O)O)(C(=O)O)O.N.[Fe+3] | |
| 264.999 | |
| Weak |
| Brown | |
| 1000 g | |
| MFCD00013099 | |
| ammonium ferric citrate, ferri seltz, ammonii ferri citras, ferric ammonium citrate, iron ammonium citrate, ammonium iron 3+ citrate, ammonium iron iii citrate, iron ammonium citrate, green, ferri-ammoniumcitrat, braunes, ferriseltz | |
| FRHBOQMZUOWXQL-UHFFFAOYSA-N | |
| azane;2-hydroxypropane-1,2,3-tricarboxylic acid;iron(3+) | |
| 118984355 | |
| Hygroscopic; Light sensitive |
Chemical Identifiers
| 1185-57-5 | |
| 264.999 | |
| FRHBOQMZUOWXQL-UHFFFAOYSA-N | |
| 118984355 | |
| C(C(=O)O)C(CC(=O)O)(C(=O)O)O.N.[Fe+3] |
| C6H11FeNO7+3 | |
| MFCD00013099 | |
| ammonium ferric citrate, ferri seltz, ammonii ferri citras, ferric ammonium citrate, iron ammonium citrate, ammonium iron 3+ citrate, ammonium iron iii citrate, iron ammonium citrate, green, ferri-ammoniumcitrat, braunes, ferriseltz | |
| azane;2-hydroxypropane-1,2,3-tricarboxylic acid;iron(3+) |
Safety and Handling
EINECSNumber : 214-686-6
RTECSNumber : GE7540000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only