missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ammonium iron(III) citrate, 16.5 to 18.5% Fe, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 612215000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Ammonium iron(III) citrate | |
Brown to Brown-Yellow | |
Powder | |
C6H11FeNO7+3 | |
15, 4044 | |
FRHBOQMZUOWXQL-UHFFFAOYSA-N | |
azane;2-hydroxypropane-1,2,3-tricarboxylic acid;iron(3+) | |
118984355 | |
16.5 to 18.5% (Fe) | |
Authentic |
16.5 to 18.5% Fe | |
1185-57-5 | |
500 g | |
MFCD00013099 | |
ammonium ferric citrate, ferri seltz, ammonii ferri citras, ferric ammonium citrate, iron ammonium citrate, ammonium iron 3+ citrate, ammonium iron iii citrate, iron ammonium citrate, green, ferri-ammoniumcitrat, braunes, ferriseltz | |
C(C(=O)O)C(CC(=O)O)(C(=O)O)O.N.[Fe+3] | |
264.999 | |
265 | |
Pass Test | |
Plastic drum |
Chemical Identifiers
1185-57-5 | |
FRHBOQMZUOWXQL-UHFFFAOYSA-N | |
118984355 | |
C(C(=O)O)C(CC(=O)O)(C(=O)O)O.N.[Fe+3] |
MFCD00013099 | |
ammonium ferric citrate, ferri seltz, ammonii ferri citras, ferric ammonium citrate, iron ammonium citrate, ammonium iron 3+ citrate, ammonium iron iii citrate, iron ammonium citrate, green, ferri-ammoniumcitrat, braunes, ferriseltz | |
azane;2-hydroxypropane-1,2,3-tricarboxylic acid;iron(3+) |
Safety and Handling
LIGHT SENSITIVE
HYGROSCOPIC
RUO – Research Use Only