missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ammonium iron(III) citrate, 16.5 to 18.5% Fe, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 211840010
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Ammonium iron(III) citrate | |
1185-57-5 | |
C6H11FeNO7 | |
15, 4044 | |
Solubility in water: 1200g/L (20 c). Other solubilities: practically insoluble in alcohol | |
C(C(=O)O)C(CC(=O)O)(C(=O)O)O.N.[Fe+3] | |
265 | |
265 | |
Authentic |
16.5 to 18.5% Fe | |
1kg | |
MFCD00013099 | |
ammonium ferric citrate, ferri seltz, ammonii ferri citras, ferric ammonium citrate, iron ammonium citrate, ammonium iron 3+ citrate, ammonium iron iii citrate, iron ammonium citrate, green, ferri-ammoniumcitrat, braunes, ferriseltz | |
FRHBOQMZUOWXQL-UHFFFAOYSA-N | |
azane;2-hydroxypropane-1,2,3-tricarboxylic acid;iron(3+) | |
118984355 | |
16.5 to 18.5% (Fe) | |
Plastic bottle |
Safety and Handling
LIGHT SENSITIVE
HYGROSCOPIC
EINECSNumber : 214-686-6
RUO â Research Use Only