Please login to your online account to display your discounted pricing

Ammonium iron(II) sulfate hexahydrate, 99+%, for analysis, ACROS Organics™


Quantity 500g
Packaging Plastic bottle

Manufacturer: fisher scientific acros  201375000

 View more versions of this product

Catalog No. AC201375000

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥99.0%
Formula Weight 392.14
Physical Form Crystals or Crystalline Powder
Quantity 500g
Color Light green
Merck Index 15, 518
Purity Grade For analysis
Phosphate 30ppm max.
Synonym Ferrous ammonium sulfate
Linear Formula (NH4)2Fe(SO4)2·6H2O
Chlorine (Cl) 10ppm max.
MDL Number MFCD00150530
Health Hazard 1 Warning
Storage Information Due to inherent oxidation, ferric ion (Fe3+) content may increase on storage
Manganese (Mn) 500ppm max.
Trace Analysis 30ppm max.
Iron (Fe3) 200ppm max.
Packaging Plastic bottle
Copper (Cu) 20ppm max.
CAS 7783-85-9
Molecular Formula H8FeN2O8S2·6H2O
Assay Percent Range 99+%
Chemical Name or Material Ammonium iron(II) sulfate hexahydrate
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Ammonium iron(II) sulfate hexahydrate
CAS 7783-85-9
Purity Grade For analysis
Assay ≥99.0%
Assay Percent Range 99+%
Molecular Formula H8FeN2O8S2·6H2O
Linear Formula (NH4)2Fe(SO4)2·6H2O
Formula Weight 392.14
MDL Number MFCD00150530
Synonym Ferrous ammonium sulfate
Accessories

Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents