Please login to your online account to display your discounted pricing

Alfa Aesar™ Ammonium iron(II) sulfate hexahydrate, 99%

$72.93 - $72.93

Chemical Identifiers

Chemical Name or Material Ammonium iron(II) sulfate hexahydrate
CAS 7783-85-9
Molecular Formula H8FeN2O8S2·6H2O
Linear Formula (NH4)2Fe(SO4)2·6H2O
Formula Weight 392.13 (284.04 Anhydrous)
MDL Number MFCD00150530
Merck Index 14,521
Synonym Iron(II) ammonium sulfate; Ferrous ammonium sulfate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1247336

  SDS
alfa aesar™
A1247336
500g Each for $72.93
Description & Specifications

Specifications

Formula Weight 392.13 (284.04 Anhydrous)
Chemical Name or Material Ammonium iron(II) sulfate hexahydrate
Synonym Iron(II) ammonium sulfate; Ferrous ammonium sulfate
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
TSCA TSCA
Odor Odorless
Assay 99%
Molecular Formula H8FeN2O8S2·6H2O
Linear Formula (NH4)2Fe(SO4)2·6H2O
MDL Number MFCD00150530
CAS 7783-85-9
Merck Index 14,521
Melting Point ≈100°C (dec.)
Quantity 500g
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number BR6500000

Solubility
Soluble in water. Insoluble in alcohol.

Notes
Air and light sensitive. Incompatible with strong acids and oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.