Please login to your online account to display your discounted pricing

Ammonium iron(II) sulfate, Dry, Acculute Standard Volumetric Solution, 0.1N, Alfa Aesar™

Air and Light Sensitive

Chemical Identifiers

Chemical Name or Material Ammonium iron(II) sulfate
CAS 7783-85-9
Physical Form Crystals
Assay Percent Range 0.1N
Molecular Formula H8FeN2O8S2
Formula Weight 284.04
MDL Number MFCD00051039
Synonym Ammonium iron(II) sulfate, hexahydrate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA88584LQ

  SDS
alfa aesar
88584LQ
1 unit CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA88584LS

  SDS
alfa aesar
88584LS
6 units CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Crystals
Formula Weight 284.04
Quantity 1 unit
Color Pale blue-green
EINECS Number 233-382-4
Odor Odorless
CAS 7783-85-9
Molecular Formula H8FeN2O8S2
Synonym Ammonium iron(II) sulfate, hexahydrate
Chemical Name or Material Ammonium iron(II) sulfate
MDL Number MFCD00051039
Assay Percent Range 0.1N
Health Hazard 1 H315-H319-H335: Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
Safety and Handling

H315-H319-H335: Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.