Please login to your online account to display your discounted pricing

Alfa Aesar™ Ammonium, Ion chromatography standard solution, Specpure™, NH4+ 1000μg/mL

Chemical Identifiers

Chemical Name or Material Ammonium Ion chromatography standard solution
Name Note NH4+ 1000μg/mL
Purity Grade Specpure™
MDL Number MFCD00143604
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA35560AE

  SDS
alfa aesar™
35560AE
100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Ammonium Ion chromatography standard solution
MDL Number MFCD00143604
Physical Form Liquid
Quantity 100mL
TSCA TSCA
Name Note NH4+ 1000μg/mL
Purity Grade Specpure™