missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ammonium iodide, ACS, 99.0% min, Alfa Aesar™

Manufacturer:  Alfa Aesar 1166822

 View more versions of this product

Catalog No. AA1166822



Specifications

Specifications

Ammonium iodide
99.0% min
MFCD00011424
ammonium iodide, azanium iodide, unii-oz8f027ldh, ammonium iodide nh4 i, oz8f027ldh, ammonium-iodid, ammonium jodatum, ammoniumiodid, hsdb 476, nh4i
UKFWSNCTAHXBQN-UHFFFAOYSA-N
azanium;iodide
25487
144.94
≤99.0%
2.514
12027-06-4
H4IN
14,529
Freely soluble in water, alcohol, glycerol, methanol
[NH4+].[I-]
144.943
CHEBI:63040
Granular
ACS
100g

Chemical Identifiers

12027-06-4
144.943
UKFWSNCTAHXBQN-UHFFFAOYSA-N
25487
azanium;iodide
H4IN
MFCD00011424
ammonium iodide, azanium iodide, unii-oz8f027ldh, ammonium iodide nh4 i, oz8f027ldh, ammonium-iodid, ammonium jodatum, ammoniumiodid, hsdb 476, nh4i
CHEBI:63040
[NH4+].[I-]
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P362-P312-P321-P405-P403+P233-P501a

Warning

TSCA : TSCA

Documents