Please login to your online account to display your discounted pricing

Ammonium iodide, 99.995% (metals basis), Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Ammonium iodide
CAS 12027-06-4
Name Note (metals basis)
Molecular Formula H4IN
Linear Formula NH4I
Formula Weight 144.94
MDL Number MFCD00011424
Merck Index 14,529
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AA4025506

  SDS
alfa aesar
4025506
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 144.94
Quantity 5g
Odor Odorless
Melting Point 551°C (Subl.)
Molecular Formula H4IN
Refractive Index 1.7031
Health Hazard 1 Warning
Chemical Name or Material Ammonium iodide
Sensitivity Light and Air Sensitive and Hygroscopic
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
CAS 12027-06-4
Mesh Size −4 Mesh
Color White to Pale Yellow
Name Note (metals basis)
TSCA TSCA
Merck Index 14,529
Assay 99.995%
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Linear Formula NH4I
MDL Number MFCD00011424
Density 2.514
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.