missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ammonium iodide, 98+%

$71.43 - $708.07

Chemical Identifiers

CAS 12027-06-4
Molecular Formula NH4I
Formula Weight 144.94
MDL Number MFCD00011424
Merck Index 14,529
Chemical Name or Material Ammonium iodide
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1249822 SDS Alfa Aesar™
A1249822
100g Each for $71.43
AAA124980E SDS Alfa Aesar™
A124980E
2500g Each for $708.07
AAA1249836 SDS Alfa Aesar™
A1249836
500g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Description

Applications
Ammonium iodide is a laboratory reagent and used as a photographic chemicals. It has wide range of applications in chemical industries.

Solubility
Soluble in water (1770 mg/ml at 25°C), methanol (400 mg/ml), glycerol (667 mg/ml), alcohol (270 mg/ml), and ethanol.

Notes
Store at 4°C. Hande under dry protective gas.
Description & Specifications

Chemical Identifiers

12027-06-4
144.94
14,529
NH4I
MFCD00011424
Ammonium iodide

Specifications

12027-06-4
144.94
14,529
551°C (subl.)
Ammonium iodide
NH4I
MFCD00011424
Odorless
100g
≥98%

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA