missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ammonium iodate, for analysis, ACROS Organics™

Chemical Identifiers

CAS 13446-09-8
Molecular Formula H4INO3
Formula Weight 192.94
MDL Number MFCD00049684
Chemical Name or Material Ammonium iodate
Assay ≥99.0%
Grade For analysis
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC390550500 SDS Thermo Fisher Scientific
AC390550500
Plastic bottle 50g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC390552500 SDS Thermo Fisher Scientific
AC390552500
Plastic bottle 250g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Description

Oxidation reagent
Description & Specifications

Chemical Identifiers

13446-09-8
192.94
Ammonium iodate
For analysis
H4INO3
MFCD00049684
≥99.0%

Specifications

13446-09-8
H4INO3
MFCD00049684
0.005% max.
0.001% max.
0.005% max.
50g
≥99.0%
Powder
192.94
5ppm max.
0.001% max.
Plastic bottle
White
Ammonium iodate
For analysis

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
May intensify fire; oxidizer.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger