missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ammonium iodate, 99%

Manufacturer:  Alfa Aesar™ 1453118

 View more versions of this product

Catalog No. AA1453118



Specifications

Specifications

Ammonium iodate
H4INO3
MFCD00049684
ammonium iodate, azanium iodate, iodic acid hio3 , ammonium salt, acmc-1bqbr, ammonium iodate 50g, iodic acid hio3 , ammonium salt 1:1
[NH4+].[O-]I(=O)=O
192.94
192.94
99%
Odorless
White
50g
13446-09-8
NH4IO3
UN1479
ZRDJERPXCFOFCP-UHFFFAOYSA-N
azanium;iodate
166805
Crystalline
3.309g/mL
Moisture Sensitive
150°C

Chemical Identifiers

13446-09-8
192.94
ZRDJERPXCFOFCP-UHFFFAOYSA-N
166805
[NH4+].[O-]I(=O)=O
H4INO3
MFCD00049684
ammonium iodate, azanium iodate, iodic acid hio3 , ammonium salt, acmc-1bqbr, ammonium iodate 50g, iodic acid hio3 , ammonium salt 1:1
azanium;iodate
Safety and Handling

Safety and Handling

GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P221-P210-P220-P305+P351+P338-P405-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Keep/Store away from clothing/combustible materials.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: OXIDIZING SOLID, N.O.S.

TSCA : TSCA

Documents