missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ammonium iodate, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 01453130

 View more versions of this product

Catalog No. AA1453130



Description

Description

Ammonium iodate is used as an oxidation reagent. Ammonium Iodate-18O3 is the labeled analog of ammonium iodate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Ammonium iodate
13446-09-8
Crystalline
H4INO3
MFCD00049684
ammonium iodate, azanium iodate, iodic acid hio3 , ammonium salt, acmc-1bqbr, ammonium iodate 50g, iodic acid hio3 , ammonium salt 1:1
ZRDJERPXCFOFCP-UHFFFAOYSA-N
azanium;iodate
166805
99%
Moisture sensitive
150°C
White
250 g
NH4IO3
UN1479
Soluble in water 20.6g/L @ 20°C
[NH4+].[O-]I(=O)=O
192.94
192.94
Odorless
3.309 g/mL

Chemical Identifiers

13446-09-8
192.94
ZRDJERPXCFOFCP-UHFFFAOYSA-N
166805
[NH4+].[O-]I(=O)=O
H4INO3
MFCD00049684
ammonium iodate, azanium iodate, iodic acid hio3 , ammonium salt, acmc-1bqbr, ammonium iodate 50g, iodic acid hio3 , ammonium salt 1:1
azanium;iodate
Safety and Handling

Safety and Handling

GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P220-P221-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c

H272-H302-H315-H319-H335

DOTInformation : Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: OXIDIZING SOLID, N.O.S.

EINECSNumber : 236-592-4

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only