Please login to your online account to display your discounted pricing

Ammonium Hydroxide , Macron Fine Chemicals™

macron

Chemical Identifiers

Chemical Name or Material Ammonium Hydroxide
Name Note (28.0-30.0% as NH 3)
CAS 1336-21-6
Molecular Formula NH4OH
Formula Weight 35.05
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MK3256212

 
avantor performance
325648
Deal-AR-Pak 6 x 2.5L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Ammonium Hydroxide
Quantity 6 x 2.5L
Name Note (28.0-30.0% as NH 3)
CAS 1336-21-6
Molecular Formula NH4OH
Formula Weight 35.05
Packaging Deal-AR-Pak