Please login to your online account to display your discounted pricing

Ammonium hydroxide, Macron Fine Chemicals™

macron

Chemical Identifiers

Chemical Name or Material Ammonium hydroxide
CAS 1336-21-6
Physical Form Liquid
Molecular Formula NH4OH
Formula Weight 35.04
Synonym Ammonia water
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

14650381

 
avantor performance
325614
500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Color Clear colorless
Formula Weight 35.04
Quantity 500mL
Chemical Name or Material Ammonium hydroxide
Density 0.91g/cm3
Molecular Formula NH4OH
CAS 1336-21-6
Synonym Ammonia water