missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ammonium Hydroxide, For Hardness, Certified, 19.2 to 20.8% (v/v), LabChem™

Supplier:  LabChem LC111604

 View more versions of this product

Catalog No. LC111604



Description

Description

Specifications

Specifications

Ammonium Hydroxide
For Hardness
94.95,5.05
H5NO
MFCD00066650
ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh
4 L
N.O
35.05
CHEBI:18219
19.2 to 20.8% (v/v)
Passes Test
Poly Bottle
Traceable to NIST
Colorless
1336-21-6,7732-18-5
as NH4OH
NH4OH
0.98g/mL
Soluble in water
VHUUQVKOLVNVRT-UHFFFAOYSA-N
amine hydrate
14923
35.05
Certified
Ammonia
0.98g/mL
Liquid

Chemical Identifiers

1336-21-6
35.05
VHUUQVKOLVNVRT-UHFFFAOYSA-N
14923
amine hydrate
H5NO
MFCD00066650
ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh
CHEBI:18219
N.O
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Toxic to aquatic life.

GHS P Statement
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
Avoid release to the environment.
If on skin: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.

Warning

Recommended Storage : Room Temperature

SDS
Documents

Documents

Promotions

Promotions