Please login to your online account to display your discounted pricing

Ammonium hydroxide, extra pure, 25% solution in water, ACROS Organics™


Quantity 10L
Packaging Plastic Jerrycan

Manufacturer: thermo fisher scientific  AC255210100

 View more versions of this product

Catalog No. AC255210100

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Chemical Name or Material Ammonium hydroxide
Color Clear colorless
Quantity 10L
CAS 1336-21-6
Health Hazard 1 Danger
Health Hazard 2 GHS P Statement
May cause respiratory irritation.
Very toxic to aquatic life.
Causes severe skin burns and eye damage.
CAS 7732-18-5
Health Hazard 3 GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chlorine (Cl) 50ppm max.
Purity Grade Extra Pure
Physical Form Liquid
Assay ≥25% (as NH3) (total base)
Density 0.9100g/mL
Molecular Formula H5NO
Linear Formula NH4OH
Formula Weight 35.05
Iron (Fe) 50ppm max.
Packaging Plastic Jerrycan
Sulfate 50ppm max.

Specifications

Chemical Name or Material Ammonium hydroxide
CAS 1336-21-6
CAS 7732-18-5
Purity Grade Extra Pure
Assay ≥25% (as NH3) (total base)
Molecular Formula H5NO
Linear Formula NH4OH
Formula Weight 35.05
Accessories

Danger

GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS P Statement
May cause respiratory irritation.
Very toxic to aquatic life.
Causes severe skin burns and eye damage.